4-Bromo-1-ethyl-1H-pyrazole 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-1-ethylpyrazole

OR59380-25G
APOSOR59380-25GEA 150 EUR
APOSOR59380-25G
4-Bromo-1-ethyl-1H-pyrazole 95+%
4-Bromo-1-ethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MW: 175,0265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235252
CAS Number: 71229-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR