N-(4-Methoxybenzyl)-N-methylbenzenesulphonamide
Supplier: Apollo Scientific
OR59400-1G
OR59400-5G
APOSOR59400-1GEA
72.7
EUR
APOSOR59400-1G
APOSOR59400-5G
N-(4-Methoxybenzyl)-N-methylbenzenesulphonamide
N-(4-Methoxybenzyl)-N-methylbenzenesulphonamide
Formula:
C₁₅H₁₇NO₃S MW: 291,365 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08070819 CAS Number: 915916-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...