2-(4-Iodophenoxy)malondialdehyde 95+%

Supplier: Apollo Scientific

OR59435-10G OR59435-1G OR59435-5G
APOSOR59435-10GEA 970 EUR
APOSOR59435-10G APOSOR59435-1G APOSOR59435-5G
2-(4-Iodophenoxy)malondialdehyde 95+%
2-(4-Iodophenoxy)malondialdehyde
Formula: C₉H₇IO₃
Storage Temperature: Ambient
MDL Number: MFCD10688277
CAS Number: 1040682-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR