4-Nitro-1H-pyrazole 98%

Supplier: Apollo Scientific
Warning

OR59445-100G OR59445-25G OR59445-500G
APOSOR59445-100GEA 60.9 EUR
APOSOR59445-100G APOSOR59445-25G APOSOR59445-500G
4-Nitro-1H-pyrazole 98%
4-Nitro-1H-pyrazole
Formula: C₃H₃N₃O₂
MW: 113,0800 g/mol
Boiling Pt: 323 °C
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00159626
CAS Number: 2075-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR