4-Methoxypiperidine

Supplier: Apollo Scientific
Warning

APOSOR59470-1GEA 49.5 EUR
APOSOR59470-1G
4-Methoxypiperidine
4-Methoxypiperidine
Formula: C₆H₁₄ClNO
MW: 115,1700 g/mol
Boiling Pt: 103 °C/113mmHg
Storage Temperature: Ambient
MDL Number: MFCD04115010
CAS Number: 4045-24-3
EINECS: 223-742-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR