(4-Methyl-1,2,5-oxadiazol-3-yl)methanol, Technical Grade
Supplier: Apollo Scientific
|
Warning
|
OR5950-100MG
OR5950-1G
APOSOR5950-100MGEA
270
EUR
APOSOR5950-100MG
APOSOR5950-1G
(4-Methyl-1,2,5-oxadiazol-3-yl)methanol, Technical Grade
(4-Methyl-1,2,5-oxadiazol-3-yl)methanol
Formula:
C₄H₆N₂O₂ Storage Temperature: Ambient |
CAS Number:
78441-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...