3-Ethynylanisole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Ethynyl-3-methoxybenzene, (3-Methoxyphenyl)ethyne, (3-Methoxyphenyl)acetylene, 1-Methoxy-3-ethynylbenzene

OR5956-1G OR5956-5G
APOSOR5956-1GEA 43.7 EUR
APOSOR5956-1G APOSOR5956-5G
3-Ethynylanisole 97%
3-Ethynylanisole
Formula: C₉H₈O
MW: 132,16 g/mol
Boiling Pt: 204…210 °C
Density: 1,04 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00160810
CAS Number: 768-70-7
UN: 3334
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR