2,5-Dimethoxycinnamic acid 99+%

Supplier: Apollo Scientific

Synonyms: 3-(2,5-Dimethoxyphenyl)acrylic acid, 2',5'-Dimethoxycinnamic acid, trans-2,5-Dimethoxycinnamic acid

OR5958-25G OR5958-5G
APOSOR5958-25GEA 69.8 EUR
APOSOR5958-25G APOSOR5958-5G
2,5-Dimethoxycinnamic acid 99+%
2,5-Dimethoxycinnamic acid
Formula: C₁₁H₁₂O₄
MW: 208,21 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004378
CAS Number: 10538-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR