2,5-Dibromoisonicotinic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2,5-Dibromopyridine-4-carboxylic acid

OR59812-1G OR59812-5G
APOSOR59812-1GEA 62.6 EUR
APOSOR59812-1G APOSOR59812-5G
2,5-Dibromoisonicotinic acid
2,5-Dibromoisonicotinic acid
Formula: C₆H₃Br₂NO₂
MW: 280,9000 g/mol
Melting Pt: 174…175 °C
Storage Temperature: Ambient
MDL Number: MFCD08741485
CAS Number: 942473-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR