2,7-Dibromo-9,9-dimethylfluorene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,7-Dibromo-9,9-dimethyl-9H-fluorene
OR5982-25G
OR5982-100G
APOSOR5982-25GEA
33.5
EUR
APOSOR5982-25G
APOSOR5982-100G
2,7-Dibromo-9,9-dimethylfluorene
2,7-Dibromo-9,9-dimethylfluorene
Formula:
C₁₅H₁₂Br₂ MW: 352.07 g/mol Melting Pt: 180…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00099471 CAS Number: 28320-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...