Oxazol-5-ylmethanamine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-(Aminomethyl)-1,3-oxazole monohydrochloride
OR59854-1G
APOSOR59854-1GEA
319
EUR
APOSOR59854-1G
Oxazol-5-ylmethanamine hydrochloride
Oxazol-5-ylmethanamine hydrochloride
Formula:
C₄H₇ClN₂O MW: 134,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06804578 CAS Number: 1196156-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...