5-Bromo-1,3,4-thiadiazol-2-amine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-5-bromo-1,3,4-thiadiazole, 1,3,4-Thiadiazol-2-amine, 5-bromo-, 5-bromo-1,3,4-thiadiazol-2-amine
OR59876-25G
OR59876-5G
APOSOR59876-25GEA
33.5
EUR
APOSOR59876-25G
APOSOR59876-5G
5-Bromo-1,3,4-thiadiazol-2-amine
5-Bromo-1,3,4-thiadiazol-2-amine
Formula:
C₂H₂BrN₃S MW: 180,02 g/mol Melting Pt: 178…182 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00464325 CAS Number: 37566-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...