Ethyl-3-amino-3-oxopropanoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl malonate monoamide, Ethyl 3-amino-3-oxopropanoate

OR59898-5G OR59898-1G
APOSOR59898-5GEA 79.9 EUR
APOSOR59898-5G APOSOR59898-1G
Ethyl-3-amino-3-oxopropanoate 98%
Ethyl-3-amino-3-oxopropanoate
Formula: C₅H₉NO₃
MW: 131,1300 g/mol
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00043150
CAS Number: 7597-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR