Diethyl ethylidenemalonate 95+%
Supplier: Apollo Scientific
Synonyms:
Diethyl 2-ethylidenemalonate
OR59915-10G
OR59915-25G
APOSOR59915-10GEA
77
EUR
APOSOR59915-10G
APOSOR59915-25G
Diethyl ethylidenemalonate 95+%
Diethyl ethylidenemalonate
Formula:
C₉H₁₄O₄ MW: 186.21 g/mol Boiling Pt: 115…118 °C/17mmHg Density: 1.019 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009145 CAS Number: 1462-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...