Ethyl isoquinoline-3-carboxylate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 3-isoquinolinecarboxylate

OR59951-1G OR59951-25G OR59951-5G
APOSOR59951-1GEA 78.6 EUR
APOSOR59951-1G APOSOR59951-25G APOSOR59951-5G
Ethyl isoquinoline-3-carboxylate 97%
Ethyl isoquinoline-3-carboxylate
Formula: C₁₂H₁₁NO₂
MW: 201,2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566077
CAS Number: 50458-79-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR