Tebuconazole 97%
Supplier: Apollo Scientific
Synonyms:
1-(4-Chlorophenyl)-4,4-dimethyl-3-(1H-1,2,4-triazol-1-ylmethyl)-3-pentanol
OR59970-5G
APOSOR59970-5GEA
37.7
EUR
APOSOR59970-5G
Tebuconazole 97%
Tebuconazole
Formula:
C₁₆H₂₂ClN₃O MW: 307,82 g/mol Melting Pt: 102,4 °C Density: 1,25 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD02674797 CAS Number: 107534-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...