4-Isopropylphenylacetic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(4-Isopropylphenyl)acetic acid

OR59979-25G
APOSOR59979-25GEA 172 EUR
APOSOR59979-25G
4-Isopropylphenylacetic acid 98%
4-Isopropylphenylacetic acid
Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Melting Pt: 51…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00041035
CAS Number: 4476-28-2
EINECS: 224-755-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR