Isophthalaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: m-Phthalaldehyde, Isophthaldicarboxaldehyde, Isophthaldialdehyde

OR59986-25G OR59986-100G OR59986-500G
APOSOR59986-25GEA 21.9 EUR
APOSOR59986-25G APOSOR59986-100G APOSOR59986-500G
Isophthalaldehyde
Isophthalaldehyde
Formula: C₈H₆O₂
MW: 134,13 g/mol
Boiling Pt: 136 °C
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00003372
CAS Number: 626-19-7
EINECS: 210-935-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR