1H-Indazole-5-carboxylic acid

Supplier: Apollo Scientific
Warning

OR60062-1G OR60062-5G
APOSOR60062-1GEA 23.2 EUR
APOSOR60062-1G APOSOR60062-5G
1H-Indazole-5-carboxylic acid
1H-Indazole-5-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 318…322 °C
Storage Temperature: Ambient
MDL Number: MFCD06804570
CAS Number: 61700-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR