5-Bromo-3-cyanoindole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-1H-indole-3-carbonitrile
OR60067-10G
OR60067-5G
APOSOR60067-10GEA
139
EUR
APOSOR60067-10G
APOSOR60067-5G
5-Bromo-3-cyanoindole 97%
5-Bromo-3-cyanoindole
Formula:
C₉H₅BrN₂ MW: 221,0500 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04127133 CAS Number: 90271-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...