4-Methoxy-3-pyridineboronic acid hydrate

Supplier: Apollo Scientific
Warning

OR60085-1G OR60085-5G
APOSOR60085-1GEA 125 EUR
APOSOR60085-1G APOSOR60085-5G
4-Methoxy-3-pyridineboronic acid hydrate
4-Methoxy-3-pyridineboronic acid hydrate
Formula: C₆H₈BNO₃·H₂O
Storage Temperature: Freezer
MDL Number: MFCD07186213
CAS Number: 1256355-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR