4-Amino-2-chlorophenol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloro-4-hydroxyaniline

OR60093-100G OR60093-25G
APOSOR60093-100GEA 227 EUR
APOSOR60093-100G APOSOR60093-25G
4-Amino-2-chlorophenol 97%
4-Amino-2-chlorophenol
Formula: C₆H₆ClNO
MW: 143,57 g/mol
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00128891
CAS Number: 3964-52-1
EINECS: 202-458-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR