4-Hydroxycyclohexanone

Supplier: Apollo Scientific
Warning

Synonyms: 4-Hydroxycyclohexanon

OR60149-25G
APOSOR60149-25GEA 142 EUR
APOSOR60149-25G
4-Hydroxycyclohexanone
4-Hydroxycyclohexanone
Formula: C₆H₁₀O₂
MW: 114,1424 g/mol
Boiling Pt: 256 °C
Melting Pt: –1 °C
Density: 1,1 g/cm³ (25 °C)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD02093817
CAS Number: 13482-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR