4-Isoxazolecarboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: Isoxazole-4-carboxylic acid

OR60164-5G
APOSOR60164-5GEA 163 EUR
APOSOR60164-5G
4-Isoxazolecarboxylic acid 98%
4-Isoxazolecarboxylic acid
Formula: C₄H₃NO₃
MW: 113,0715 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD06738806
CAS Number: 6436-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR