Tetrahydropyran-2-ylmethylamine 95%
Supplier: Apollo Scientific
|
Danger
|
OR60205-1G
OR60205-250MG
OR60205-5G
APOSOR60205-1GEA
205
EUR
APOSOR60205-1G
APOSOR60205-250MG
APOSOR60205-5G
Tetrahydropyran-2-ylmethylamine 95%
Tetrahydropyran-2-ylmethylamine
Formula:
C₆H₁₃NO Boiling Pt: 42 °C/0,1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD06738971 CAS Number: 6628-83-7 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...