2,5-Dimethylpyrazine 98+%
Supplier: Apollo Scientific
|
Warning
|
OR6021-100G
OR6021-1KG
OR6021-250G
APOSOR6021-100GEA
118
EUR
APOSOR6021-100G
APOSOR6021-1KG
APOSOR6021-250G
2,5-Dimethylpyrazine 98+%
2,5-Dimethylpyrazine
Formula:
C₆H₈N₂ MW: 108,14 g/mol Boiling Pt: 155 °C Melting Pt: 15 °C Density: 0,99 g/cm³ (25 °C) Flash Pt: 63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006147 CAS Number: 123-32-0 EINECS: 204-618-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...