2-Chloro-6-hydrazinopyrazine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-6-hydrazinylpyrazine

OR60236-1G
APOSOR60236-1GEA 56.7 EUR
APOSOR60236-1G
2-Chloro-6-hydrazinopyrazine
2-Chloro-6-hydrazinopyrazine
Formula: C₄H₅ClN₄
MW: 144,56 g/mol
Melting Pt: 136…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09049072
CAS Number: 63286-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR