2-Ethylfuran

Supplier: Apollo Scientific

OR6028-10G OR6028-25G
APOSOR6028-10GEA 49.5 EUR
APOSOR6028-10G APOSOR6028-25G
2-Ethylfuran
2-Ethylfuran
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 92…93 °C/768mmHg
Density: 0.912 g/cm³ (25 °C)
Flash Pt: –2 °C
Storage Temperature: Ambient
MDL Number: MFCD00003259
CAS Number: 3208-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR