5-[2-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole 97%
Supplier: Apollo Scientific
|
Danger
|
OR6068-1G
OR6068-250MG
APOSOR6068-1GEA
396
EUR
APOSOR6068-1G
APOSOR6068-250MG
5-[2-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole 97%
5-[2-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
Formula:
C₁₀H₉BrN₂O Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD09064955 CAS Number: 892501-91-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...