3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride

Supplier: Apollo Scientific
Warning

Synonyms: 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride

OR61012-1G OR61012-5G OR61012-10G
APOSOR61012-1GEA 71.3 EUR
APOSOR61012-1G APOSOR61012-5G APOSOR61012-10G
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Formula: C₁₃H₁₆ClNOS
MW: 269,79 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00011959
CAS Number: 4568-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR