2-Amino-5-phenylpyrazine 98%
Supplier: Apollo Scientific
|
Warning
|
OR61038-10G
OR61038-1G
OR61038-5G
APOSOR61038-10GEA
793
EUR
APOSOR61038-10G
APOSOR61038-1G
APOSOR61038-5G
2-Amino-5-phenylpyrazine 98%
2-Amino-5-phenylpyrazine
Formula:
C₁₀H₉N₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD01646266 CAS Number: 13535-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...