(1-Aminocyclohexyl)methanol 98%

Supplier: Apollo Scientific
Warning

OR61050-250MG
APOSOR61050-250MGEA 50.9 EUR
APOSOR61050-250MG
(1-Aminocyclohexyl)methanol 98%
(1-Aminocyclohexyl)methanol
Formula: C₇H₁₅NO
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD12198034
CAS Number: 4313-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR