Ethyl-2-(4-isobutylphenyl)propionate 98%
Supplier: Apollo Scientific
|
Warning
|
OR61054-1G
OR61054-250MG
APOSOR61054-1GEA
87
EUR
APOSOR61054-1G
APOSOR61054-250MG
Ethyl-2-(4-isobutylphenyl)propionate 98%
Ethyl-2-(4-isobutylphenyl)propionate
Formula:
C₁₅H₂₂O₂ MW: 234,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00869901 CAS Number: 41283-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...