1-Benzylpiperidine-4-carbaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
N-Benzylpiperidine-4-carboxaldehyde, 1-Benzyl-4-piperidinecarboxaldehyde
OR61172-25G
APOSOR61172-25GEA
77
EUR
APOSOR61172-25G
1-Benzylpiperidine-4-carbaldehyde 95%
1-Benzylpiperidine-4-carbaldehyde
Formula:
C₁₃H₁₇NO MW: 203,28 g/mol Boiling Pt: 315 °C Density: 1,026 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD02258902 CAS Number: 22065-85-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...