N-Carbobenzyloxy-L-alanine 97%

Supplier: Apollo Scientific
Warning

Synonyms: N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine

OR61272-100G OR61272-1KG OR61272-500G
APOSOR61272-100GEA 61.1 EUR
APOSOR61272-100G APOSOR61272-1KG APOSOR61272-500G
N-Carbobenzyloxy-L-alanine 97%
N-Carbobenzyloxy-L-alanine
Formula: C₁₁H₁₃NO₄
MW: 223,23 g/mol
Melting Pt: 82…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002640
CAS Number: 1142-20-7
EINECS: 214-532-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR