2-Butene 99%
Supplier: Apollo Scientific
|
Danger
|
OR61316-200G
OR61316-400G
OR61316-100G
OR61316-250G
APOSOR61316-200GEA
1210
EUR
APOSOR61316-200G
APOSOR61316-400G
APOSOR61316-100G
APOSOR61316-250G
2-Butene 99%
2-Butene
Formula:
C₄H₈ Boiling Pt: 1 °C Melting Pt: –140 °C Density: 0,626 g/cm³ (25 °C) Flash Pt: –34 °C Storage Temperature: Ambient |
CAS Number:
107-01-7 EINECS: 203-452-9 UN: 1012 ADR: 2.1, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...