3-Amino-1-Boc-piperidine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

OR6132-1G OR6132-5G
APOSOR6132-1GEA 14.7 EUR
APOSOR6132-1G APOSOR6132-5G
3-Amino-1-Boc-piperidine 97%
3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861219
CAS Number: 184637-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR