3-Iodo-5-nitrobenzoic acid 97%
Supplier: Apollo Scientific
|
Warning
|
OR61357-1G
OR61357-25G
OR61357-5G
APOSOR61357-1GEA
66.9
EUR
APOSOR61357-1G
APOSOR61357-25G
APOSOR61357-5G
3-Iodo-5-nitrobenzoic acid 97%
3-Iodo-5-nitrobenzoic acid
Formula:
C₇H₄INO₄ Melting Pt: 166…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068827 CAS Number: 6313-17-3 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...