Diethyl-4-chloro-3,5-pyridinedicarboxylate 95%

Supplier: Apollo Scientific
Warning

OR61379-1G OR61379-500MG OR61379-5G
APOSOR61379-1GEA 306 EUR
APOSOR61379-1G APOSOR61379-500MG APOSOR61379-5G
Diethyl-4-chloro-3,5-pyridinedicarboxylate 95%
Diethyl-4-chloro-3,5-pyridinedicarboxylate
Formula: C₁₁H₁₂ClNO₄
MW: 257,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025855
CAS Number: 244638-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR