(S)-N-Boc-2-hydroxymethylmorpholine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(S)-tert-Butyl 2-(hydroxymethyl)morpholine-4-carboxylate, (2S)-2-(Hydroxymethyl)morpholine, N-BOC protected
OR61432-1G
APOSOR61432-1GEA
21.9
EUR
APOSOR61432-1G
(S)-N-Boc-2-hydroxymethylmorpholine 97%
(S)-N-Boc-2-hydroxymethylmorpholine
Formula:
C₁₀H₁₉NO₄ MW: 217,265 g/mol Melting Pt: 60…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD09260606 CAS Number: 135065-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...