cis-4-Amino-1-methylcyclohexanol 97%

Supplier: Apollo Scientific
Danger

Synonyms: cis--4-Amino-1-methylcyclohexanol

OR61449-1G OR61449-5G
APOSOR61449-1GEA 163 EUR
APOSOR61449-1G APOSOR61449-5G
cis-4-Amino-1-methylcyclohexanol 97%
cis-4-Amino-1-methylcyclohexanol
Formula: C₇H₁₅NO
MW: 129.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18791201
CAS Number: 177906-46-6
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR