Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
OR6145-10G
OR6145-1G
OR6145-5G
APOSOR6145-10GEA
84.4
EUR
APOSOR6145-10G
APOSOR6145-1G
APOSOR6145-5G
Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate
Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
Formula:
C₉H₁₃NO₂S MW: 199,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085051 CAS Number: 4815-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...