Pinacol
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2,3-Dimethylbutane-2,3-diol, Tetramethylethylene glycol, 2,3-Dimethyl-2,3-butanediol
OR61505-100G
OR61505-25G
OR61505-500G
APOSOR61505-100GEA
34.8
EUR
APOSOR61505-100G
APOSOR61505-25G
APOSOR61505-500G
Pinacol
Pinacol
Formula:
C₆H₁₄O₂ MW: 118,18 g/mol Boiling Pt: 174 °C Melting Pt: 40…43 °C Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004462 CAS Number: 76-09-5 EINECS: 200-933-5 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...