3-(Pyrimidin-2-yl)benzaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
OR6164-1G
OR6164-250MG
OR6164-5G
APOSOR6164-1GEA
256
EUR
APOSOR6164-1G
APOSOR6164-250MG
APOSOR6164-5G
3-(Pyrimidin-2-yl)benzaldehyde 95%
3-(Pyrimidin-2-yl)benzaldehyde
Formula:
C₁₁H₈N₂O Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD08056283 CAS Number: 263349-22-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...