4-Aminomethylbenzyl alcohol 95%
Supplier: Apollo Scientific
|
Danger
|
OR6193-1G
OR6193-250MG
OR6193-5G
APOSOR6193-1GEA
58
EUR
APOSOR6193-1G
APOSOR6193-250MG
APOSOR6193-5G
4-Aminomethylbenzyl alcohol 95%
4-Aminomethylbenzyl alcohol
Formula:
C₈H₁₁NO MW: 137,1790 g/mol Melting Pt: 82…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD06213766 CAS Number: 39895-56-2 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...