2-(Cyanomethyl)benzoic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Carboxyphenylacetonitrile

OR6244-1G OR6244-250MG
APOSOR6244-1GEA 183 EUR
APOSOR6244-1G APOSOR6244-250MG
2-(Cyanomethyl)benzoic acid 97%
2-(Cyanomethyl)benzoic acid
Formula: C₉H₇NO₂
MW: 161,16 g/mol
Storage Temperature: Ambient
CAS Number: 6627-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR