5-(4-Chlorophenyl)-4-oxazolecarboxylic acid 98%

Supplier: Apollo Scientific
Warning

OR6278-100MG OR6278-1G
APOSOR6278-100MGEA 58.2 EUR
APOSOR6278-100MG APOSOR6278-1G
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid 98%
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid
Formula: C₁₀H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00105450
CAS Number: 143659-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR