2-Phenyl-1H-imidazole-4-carboxylic acid hydrate 95%

Supplier: Apollo Scientific
Warning

OR6287-250MG OR6287-100MG
APOSOR6287-250MGEA 75.7 EUR
APOSOR6287-250MG APOSOR6287-100MG
2-Phenyl-1H-imidazole-4-carboxylic acid hydrate 95%
2-Phenyl-1H-imidazole-4-carboxylic acid hydrate
Formula: C₁₀H₁₀N₂O₃
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD04974056
CAS Number: 77498-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR