6-Piperidin-1-ylpyridazine-3-carboxylic acid 97%
Supplier: Apollo Scientific
|
Warning
|
OR6300-1G
OR6300-250MG
APOSOR6300-1GEA
377
EUR
APOSOR6300-1G
APOSOR6300-250MG
6-Piperidin-1-ylpyridazine-3-carboxylic acid 97%
6-Piperidin-1-ylpyridazine-3-carboxylic acid
Formula:
C₁₀H₁₃N₃O₂ MW: 207,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08235212 CAS Number: 914637-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...