3,5-Dimethylbenzylamine 97%
Supplier: Apollo Scientific
|
Warning
|
OR6321-10G
OR6321-2G
OR6321-5G
OR6321-1G
OR6321-25G
APOSOR6321-10GEA
280
EUR
APOSOR6321-10G
APOSOR6321-2G
APOSOR6321-5G
APOSOR6321-1G
APOSOR6321-25G
3,5-Dimethylbenzylamine 97%
3,5-Dimethylbenzylamine
Formula:
C₉H₁₃N Boiling Pt: 102…104 °C/10mmHg Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040753 CAS Number: 78710-55-1 UN: 2924 ADR: 3+8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...